Learn More
Tocris Bioscience™ Pomalidomide 5'-piperidine-acid
Description
Pomalidomide 5'-piperidine-acid is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a piperidine linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 1-[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]-4-piperidinecarboxylic acid |
| CAS | 2467965-70-2 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C19H19N3O6 |
| Purity | 0.95 |