missing translation for 'onlineSavingsMsg'
Learn More

Piperidine, Sigma-Aldrich™

Product Code. 15871771
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871771 2.5 L 2.5 L
15851771 100 mL 100mL
15861771 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15871771 Supplier Sigma Aldrich Supplier No. 8222992500
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

For synthesis

Piperidine is a heterocyclic amine with a six-membered ring containing five methylene bridges (-CH2-) and one amine bridge (-NH-). Numerous alkaloids, pharmaceuticals, agrochemicals, and synthetic and biological intermediates contain the piperidine moiety. The commercial applications of piperidine include: solvent for curing agents and epoxy resins. In organic synthesis, it serves as an intermediate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-89-4

Specifications

Melting Point -10.8°C
Density 0.86 g/cm3 at 20°C
Boiling Point 106°C/1013 hPa
MDL Number MFCD00005979
Quantity 2.5 L
Synonym Piperidine, Hexahydropyridine, Pentamethyleneimine, Azacyclohexane, Azinane, Piperidine
Molecular Weight (g/mol) 85.15
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.