CAS RN 110-89-4
Piperidine, 99%, extra pure
CAS: 110-89-4 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 InChI Key: NQRYJNQNLNOLGT-UHFFFAOYSA-N IUPAC Name: piperidine SMILES: C1CCNCC1
Thermo Scientific Chemicals Piperidine, 99%
CAS: 110-89-4 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 InChI Key: NQRYJNQNLNOLGT-UHFFFAOYSA-N IUPAC Name: piperidine SMILES: C1CCNCC1
Piperidine, 99.5%, purified by redistillation, AcroSeal™
CAS: 110-89-4 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 InChI Key: NQRYJNQNLNOLGT-UHFFFAOYSA-N IUPAC Name: piperidine SMILES: C1CCNCC1
Piperidine, Sigma-Aldrich™
CAS: 110-89-4
Piperidine, Pure, Fisher Chemical™
CAS: 110-89-4 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 MDL Number: 5979 InChI Key: NQRYJNQNLNOLGT-UHFFFAOYSA-N Synonym: hexahydropyridine,azacyclohexane,cyclopentimine,cypentil,hexazane,pentamethyleneimine,piperidin,pentamethylenimine,perhydropyridine,pentamethyleneamine PubChem CID: 8082 ChEBI: CHEBI:18049 IUPAC Name: piperidine SMILES: C1CCNCC1
Piperidine, ≥99.5%, Solstice
CAS: 110-89-4 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 MDL Number: MFCD00005979 InChI Key: NQRYJNQNLNOLGT-UHFFFAOYSA-N Synonym: hexahydropyridine,azacyclohexane,cyclopentimine,cypentil,hexazane,pentamethyleneimine,piperidin,pentamethylenimine,perhydropyridine,pentamethyleneamine PubChem CID: 8082 ChEBI: CHEBI:18049 IUPAC Name: piperidine SMILES: C1CCNCC1
Piperidine, ≥99.0% (GC/T), Honeywell™
CAS: 110-89-4 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 MDL Number: MFCD00005979 InChI Key: NQRYJNQNLNOLGT-UHFFFAOYSA-N Synonym: hexahydropyridine,azacyclohexane,cyclopentimine,cypentil,hexazane,pentamethyleneimine,piperidin,pentamethylenimine,perhydropyridine,pentamethyleneamine PubChem CID: 8082 ChEBI: CHEBI:18049 IUPAC Name: piperidine SMILES: C1CCNCC1