Learn More
Piperidine, ≥99.0% (GC/T), Honeywell™
Product Code. 15687160
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15687160 1 L 1 L
15697160 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15687160 Supplier Honeywell Riedel-de Haën Supplier No. 806401L
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Currently Unavailable: We have no information when this product will be available.
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Chemical Identifiers
CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00005979
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
Synonym hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine
PubChem CID 8082
ChEBI CHEBI:18049
IUPAC Name piperidine
SMILES C1CCNCC1
Specifications
Density 0.862g/mL at 20°C
Flash Point 16°C (60.8°F)
Quantity 1 L
UN Number UN2401
Beilstein 102438
Formula Weight 85.15g/mol
Percent Purity ≥99.0% (GC/T)
Chemical Name or Material Piperidine