missing translation for 'onlineSavingsMsg'
Learn More

Pilaralisib, TRC

Product Code. 30422037
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422037 5 mg 5mg
30419694 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422037 Supplier Toronto Research Chemicals Supplier No. TRCP4413905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934526-89-3
Molecular Formula C25H25ClN6O4S
Molecular Weight (g/mol) 541.02
Synonym 2-amino-N-(3-(N-(3-((2-Chloro-5-methoxyphenyl)amino)quinoxalin-2-yl)sulfamoyl)phenyl)-2-methylpropanamide
IUPAC Name 2-amino-N-[3-[[3-(2-chloro-5-methoxyanilino)quinoxalin-2-yl]sulfamoyl]phenyl]-2-methylpropanamide
SMILES COc1ccc(Cl)c(Nc2nc3ccccc3nc2NS(=O)(=O)c4cccc(NC(=O)C(C)(C)N)c4)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C25H25ClN6O4S/c1-25(2,27)24(33)28-15-7-6-8-17(13-15)37(34,35)32-23-22(29-19-9-4-5-10-20(19)30-23)31-21-14-16(36-3)11-12-18(21)26/h4-14H,27H2,1-3H3,(H,28,33)(H,29,31)(H,30,32)
Formula Weight 540.1347
Chemical Name or Material Pilaralisib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.