CAS RN 934526-89-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Pilaralisib, TRC
CAS: 934526-89-3 Molecular Formula: C25H25ClN6O4S Molecular Weight (g/mol): 541.02 Synonym: 2-amino-N-(3-(N-(3-((2-Chloro-5-methoxyphenyl)amino)quinoxalin-2-yl)sulfamoyl)phenyl)-2-methylpropanamide IUPAC Name: 2-amino-N-[3-[[3-(2-chloro-5-methoxyanilino)quinoxalin-2-yl]sulfamoyl]phenyl]-2-methylpropanamide SMILES: COc1ccc(Cl)c(Nc2nc3ccccc3nc2NS(=O)(=O)c4cccc(NC(=O)C(C)(C)N)c4)c1
2
Pilaralisib, MedChemExpress
MedChemExpress Pilaralisib (XL147; SAR245408) is a potent and highly selective class I PI3Ks inhibitor with IC50s of 39 nM, 383 nM, 23 nM and 36 nM for PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ.