Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Product Code. 12201203 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 Columns
5 Columns
Unit Size:
Pack of 25
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12201203 25 Columns Pack of 25
12391973 5 Columns Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12201203 Supplier Thermo Scientific™ Supplier No. 88277
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.
Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1 hour; with this spin cup format, protein recovery is ≥85%.
Features of High Capacity Endotoxin Removal Resin Spin Columns:
  • High capacity—bind up to 2,000,000 EU/mL† to eliminate >99% of endotoxins
  • Selective—recover ≥85% of your protein sample
  • High performance—meets FDA guidelines by reducing final EU concentration to <5 EU/mL
  • Fast—our new spin cup format enables endotoxin depletion within 1 hour
  • Clean—single-use spin columns avoid cross contamination of samples
  • Optimized—spin cups are optimized for different sample volumes
The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(ε-lysine), as an affinity ligand to selectively bind endotoxins. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers.
Requires:
  • Endotoxin-free water
Applications:
  • Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.
Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This spin cup format is a fast, single-use method to remove endotoxins from protein samples in as little as 1 hour. The spin cups use a batch format to bind and remove endotoxins while allowing for >85% protein recovery.
† One endotoxin unit/mL (EU/mL) equals approximately 0.1ng endotoxin/mL of solution.
Related Products
  • Pierce™ High Capacity Endotoxin Removal Resin (Cat. No. 88270)
  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL (Cat. No. 88273)
  • Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL (Cat. No. 88274)
TRUSTED_SUSTAINABILITY
Specifications
Formulation 22 mL-capacity centrifuge columns containing 1 mL resin
Content And Storage Upon receipt store at 4°C.
Form Liquid
Product Line Pierce
Quantity 25 Columns
Target Endotoxin Removal
Type Spin Column
Stationary Phase Proprietary Ligand
Format Spin Column
Column Type Affinity, Cellulose Resin
Capacity (Metric) Up to 2,000,000 EU/mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pierce High Capacity Endotoxin Removal Spin Columns, 1 mL
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol