missing translation for 'onlineSavingsMsg'
Learn More

Phthaloyl Dichloride, Sigma-Aldrich™

Product Code. 15841341
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841341 250 mL 250mL
15831341 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15841341 Supplier Sigma Aldrich Supplier No. 8210290250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-95-9

Specifications

Melting Point 16°C
Density 1.41 g/cm3 at 20°C
Molecular Formula C6H4-1,2-(Cocl)2
MDL Number MFCD00000666
Quantity 250 mL
Synonym Phthaloyl dichloride, Phthalic acid dichloride, Phthaloyl dichloride
Molecular Weight (g/mol) 203.02
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.