accessibility menu, dialog, popup

UserName

CAS RN 88-95-9

ClOClO

CAS RN 88-95-9

IUPAC Name: benzene-1,2-dicarbonyl dichloride
Synonyms: phthaloyl chloride
Molecular Weight (g/mol): 203.02
Molecular Formula: C8H4Cl2O2
InChi Key: FYXKZNLBZKRYSS-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC=C1C(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)