Learn More
Phosphatase buffer 1 (5X), Thermo Scientific™
Product Code. 15474669
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. Quantity unitSize
15474669 250 mL 250mL
15464669 100 mL 100mL
2 options
Product Code. 15474669
Brand: Thermo Scientific Alfa Aesar J62267.AK
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Formulation: 100mM Tris (pH 7.5) and 100mM magnesium chloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phosphatase buffer 1 (5X)
Physical Form Liquid
Quantity 250 mL
Formulation 100mM Tris (pH 7.5) and 100mM magnesium chloride
For Research Use Only