missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Phorbol-12-myristate-13-acetate, >99%

Product Code. 11417082 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417082 25 mg 25mg
11407082 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417082 Supplier MP Biomedicals™ Supplier No. 0215186425
  • ONLINE EXCLUSIVE PRICE
    1620.00 EUR
    1542.45 EUR / 25mg
    Save 77.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phorbol-12-myristate-13-acetate, >99% is a potent tumor promoter and activates protein kinase C in vivo and in vitro.

  • PMA′s effects on PKC are attributed to its similarity to diacylgylerol, a natural activator of PKC.
  • PMA activates Ca2+− ATPase and potentiates forskolin-induced cAMP formation.
  • It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

Specifications

Purity >98% (HPLC)
For Use With (Application) Potent tumor promoter, Activates protein kinase C in vivo and in vitro
Solubility Practically insoluble.
Molecular Formula C36H56O8
CAS 16561-29-8
Content And Storage Store at − 20°C. Protect from light.
Quantity 25 mg
Molecular Weight (g/mol) 616.84 g/mol
Melting Point 162°F
Vapor Pressure 1.8X10-17 mm Hg at 25°C
Form Solid
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.