Learn More
(Phenylsulfonyl)acetamide, 98+%, Thermo Scientific™
Product Code. 11343346
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343346 25 g 25g
11333346 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11343346 Supplier Thermo Scientific Alfa Aesar Supplier No. L12741.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35008-50-5
Molecular Formula C8H9NO3S
Molecular Weight (g/mol) 199.22
MDL Number MFCD00041320
InChI Key LKYNWTGTRVZBPA-UHFFFAOYSA-N
Synonym 2-benzenesulfonyl acetamide, 2-phenylsulfonyl acetamide, phenylsulfonyl acetamide, phenylsulfonylacetamide, acmc-20aplb, enamine_000243, phenylsulphonyl acetamide, alpha-phenylsulfonylacetamide, 2-phenylsulfonyl ethanamide, acetamide, 2-phenylsulfonyl
PubChem CID 1810305
IUPAC Name 2-(benzenesulfonyl)acetamide
SMILES NC(=O)CS(=O)(=O)C1=CC=CC=C1
Specifications
Melting Point 154°C to 158°C
Quantity 25 g
Beilstein 2101624
Formula Weight 199.23
Percent Purity ≥98%
Chemical Name or Material (Phenylsulfonyl)acetamide