accessibility menu, dialog, popup

UserName

SOOONH2

CAS RN 35008-50-5

IUPAC Name: 2-(benzenesulfonyl)acetamide
Synonyms: 2-(benzenesulfonyl)acetamide
Molecular Weight (g/mol): 199.22
Molecular Formula: C8H9NO3S
InChi Key: LKYNWTGTRVZBPA-UHFFFAOYSA-N
SMILES: NC(=O)CS(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results