missing translation for 'onlineSavingsMsg'
Learn More

Phenylethyl 3-methylcaffeate, Thermo Scientific™

Product Code. p-7042971
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496539 50 mg 50mg
15486539 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496539 Supplier Thermo Scientific Alfa Aesar Supplier No. J64722.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71835-85-3
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.338
MDL Number MFCD18385026
InChI Key CZQNYPBIOHVQQN-CSKARUKUSA-N
Synonym phenylethyl 3-methylcaffeate, phenyethyl 3-methylcaffeate, phenethyl ferulate, ccris 7791, caffeic acid 3-methyl phenethyl ester, phenylethyl-3-methylcaffeate, 2-phenylethyl 3-4-hydroxy-3-methoxyphenyl-2-propenoate, 2-phenylethyl 2e-3-4-hydroxy-3-methyloxy phenyl prop-2-enoate, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl-, 2-phenylethyl ester, phenetyl ferulate
PubChem CID 5284444
IUPAC Name 2-phenylethyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES COC1=C(C=CC(=C1)C=CC(=O)OCCC2=CC=CC=C2)O

Specifications

Melting Point 80°C to 81°C
Color White
Quantity 50 mg
Formula Weight 298.33
Physical Form Solid
Chemical Name or Material Phenylethyl 3-methylcaffeate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.