accessibility menu, dialog, popup

UserName

CAS RN 71835-85-3

IUPAC Name: 2-phenylethyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Synonyms: 2-phenylethyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 298.34
Molecular Formula: C18H18O4
InChi Key: CZQNYPBIOHVQQN-CSKARUKUSA-N
SMILES: COC1=CC(\C=C\C(=O)OCCC2=CC=CC=C2)=CC=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results