Description
The stand has a clear polyacryl base in a standard SBS 96-well microplate footprint. The use of 24 extremely strong permanent magnets enables easy and fast magnetic separation in as little as 30 seconds. It sits flat on a lab bench as well as in a microplate adapter on robotic workstations.
Specifications
Specifications
| Shipping Condition | Room Temperature |
| Product Line | KingFisher™ Flex |
| Quantity | 1 Stand |
| Type | Magnetic Stand |