missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin, bacteria-like protein Antibody (V71), DyLight 350, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Peroxiredoxin, bacteria-like protein Monoclonal specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Specifications
Specifications
| Antigen | Peroxiredoxin, bacteria-like protein |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | V71 |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Host Species | Mouse |
| Immunogen | Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite). |
| Purification Method | Protein A purified |
| Show More |
For Research Use Only (RUO)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?