missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctyltriethoxysilane, TRC

Product Code. 30179156
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179156 250 mg 250mg
30179006 50 mg 50mg
30179025 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179156 Supplier Toronto Research Chemicals Supplier No. TRCP596230250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96305-13-4
Molecular Formula C14H15F17O3Si
Molecular Weight (g/mol) 582.33
Synonym Triethoxy(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)silane, Perflurooctyltriethoxysilane, Triethoxy(heptadecafluorooctyl)silane
IUPAC Name triethoxy(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)silane
SMILES CCO[Si](OCC)(OCC)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H15F17O3Si/c1-4-32-35(33-5-2,34-6-3)14(30,31)12(25,26)10(21,22)8(17,18)7(15,16)9(19,20)11(23,24)13(27,28)29/h4-6H2,1-3H3
Formula Weight 582.0519
Chemical Name or Material Perfluorooctyltriethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.