CAS RN 96305-13-4
CAS RN 96305-13-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Perfluorooctyltriethoxysilane, TRC
CAS: 96305-13-4 Molecular Formula: C14H15F17O3Si Molecular Weight (g/mol): 582.33 Synonym: Triethoxy(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)silane,Perflurooctyltriethoxysilane,Triethoxy(heptadecafluorooctyl)silane IUPAC Name: triethoxy(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)silane SMILES: CCO[Si](OCC)(OCC)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F