missing translation for 'onlineSavingsMsg'
Learn More

Perfluorononan-1-oic-acid Ammonium Salts, TRC

Product Code. 18060650
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060650 500 mg 500mg
18014050 250 mg 250mg
18082040 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18060650 Supplier Toronto Research Chemicals Supplier No. TRCP286555500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4149-60-4
Molecular Formula C9 F17 O2 . H4 N
Molecular Weight (g/mol) 481.106
Synonym Heptadecafluoro-nonanoic Acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluorononanoic Acid, C 1800, Heptadecafluorononanoic Acid, PFNA, Perfluorononanoic Acid, Perfluoropelargonic Acid, Nonanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, ammonium salt (1:1), Nonanoic acid, heptadecafluoro-, ammonium, salt (8CI,9CI), Ammonium perfluorononanoate, Ammonium perfluoropelargonate
IUPAC Name ammonium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
SMILES [NH4+].[O-]C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9HF17O2.H3N/c10-2(11,1(27)28)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)26;/h(H,27,28);1H3
Formula Weight 480.9971
Percent Purity >95%
Chemical Name or Material Perfluorononan-1-oic-acid Ammonium Salts
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.