CAS RN 4149-60-4
CAS RN 4149-60-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Perfluorononan-1-oic-acid Ammonium Salts, TRC
CAS: 4149-60-4 Molecular Formula: C9 F17 O2 . H4 N Molecular Weight (g/mol): 481.106 Synonym: Heptadecafluoro-nonanoic Acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluorononanoic Acid,C 1800,Heptadecafluorononanoic Acid,PFNA,Perfluorononanoic Acid,Perfluoropelargonic Acid,Nonanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, ammonium salt (1:1),Nonanoic acid, heptadecafluoro-, ammonium,salt (8CI,9CI),Ammonium perfluorononanoate,Ammonium perfluoropelargonate IUPAC Name: ammonium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate SMILES: [NH4+].[O-]C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F