Learn More
Pentachloroacetone, TRC
Product Code. 30135410
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135410 1 g 1g
30004225 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135410 Supplier Toronto Research Chemicals Supplier No. TRCP2379001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1768-31-6
Molecular Formula C3 H Cl5 O
Molecular Weight (g/mol) 230.3
Synonym 1,1,1,3,3-Pentachloro-2-propanone, Pentachloro-2-propanone, 1,1,1,3,3-Pentachloropropanone;
IUPAC Name 1,1,1,3,3-pentachloropropan-2-one
SMILES ClC(Cl)C(=O)C(Cl)(Cl)Cl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C3HCl5O/c4-2(5)1(9)3(6,7)8/h2H
Formula Weight 227.847
Chemical Name or Material Pentachloroacetone