CAS RN 1768-31-6
CAS RN 1768-31-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pentachloroacetone, TRC
CAS: 1768-31-6 Molecular Formula: C3 H Cl5 O Molecular Weight (g/mol): 230.3 Synonym: 1,1,1,3,3-Pentachloro-2-propanone,Pentachloro-2-propanone,1,1,1,3,3-Pentachloropropanone; IUPAC Name: 1,1,1,3,3-pentachloropropan-2-one SMILES: ClC(Cl)C(=O)C(Cl)(Cl)Cl