missing translation for 'onlineSavingsMsg'
Learn More

Pencycuron, TRC

Product Code. 30138283
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138283 1 g 1g
30137406 10 g 10g
17961275 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138283 Supplier Toronto Research Chemicals Supplier No. TRCP2216001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66063-05-6
Molecular Formula C19 H21 Cl N2 O
Molecular Weight (g/mol) 328.84
Synonym N-[(4-Chlorophenyl)methyl]-N-cyclopentyl-N'-phenylurea, Monceraen 250SC, Monceren, NTN 19701, Pencycuron
IUPAC Name 1-[(4-chlorophenyl)methyl]-1-cyclopentyl-3-phenylurea
SMILES Clc1ccc(CN(C2CCCC2)C(=O)Nc3ccccc3)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H21ClN2O/c20-16-12-10-15(11-13-16)14-22(18-8-4-5-9-18)19(23)21-17-6-2-1-3-7-17/h1-3,6-7,10-13,18H,4-5,8-9,14H2,(H,21,23)
Formula Weight 328.1342
Chemical Name or Material Pencycuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.