CAS RN 66063-05-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Pencycuron, TRC
CAS: 66063-05-6 Molecular Formula: C19 H21 Cl N2 O Molecular Weight (g/mol): 328.84 Synonym: N-[(4-Chlorophenyl)methyl]-N-cyclopentyl-N'-phenylurea,Monceraen 250SC,Monceren,NTN 19701,Pencycuron IUPAC Name: 1-[(4-chlorophenyl)methyl]-1-cyclopentyl-3-phenylurea SMILES: Clc1ccc(CN(C2CCCC2)C(=O)Nc3ccccc3)cc1
2
Pencycuron 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality