missing translation for 'onlineSavingsMsg'
Learn More

Penconazole, TRC

Product Code. 30136889
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136889 5 g 5g
17923885 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136889 Supplier Toronto Research Chemicals Supplier No. TRCP2218005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66246-88-6
Molecular Formula C13 H15 Cl2 N3
Molecular Weight (g/mol) 284.18
Synonym 1H-1,2,4-Triazole, 1-[2-(2,4-dichlorophenyl)pentyl]-, 1-[2-(2,4-Dichlorophenyl)pentyl]-1H-1,2,4-triazole, (±)-Penconazole, CGA 71818, Erluejunzuo, Ofir, Omnex, Omnex WP 10, Penconazole, Pine gel, Regal gel, Topas, Topas (fungicide), Topaze
IUPAC Name 1-[2-(2,4-dichlorophenyl)pentyl]-1,2,4-triazole
SMILES CCCC(Cn1cncn1)c2ccc(Cl)cc2Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H15Cl2N3/c1-2-3-10(7-18-9-16-8-17-18)12-5-4-11(14)6-13(12)15/h4-6,8-10H,2-3,7H2,1H3
Formula Weight 283.0643
Chemical Name or Material Penconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.