CAS RN 66246-88-6
IUPAC Name:
1-[2-(2,4-dichlorophenyl)pentyl]-1H-1,2,4-triazole
Synonyms:
penconazole
Molecular Weight (g/mol):
284.18
Molecular Formula:
C13H15Cl2N3
InChi Key:
WKBPZYKAUNRMKP-UHFFFAOYNA-N
SMILES:
CCCC(CN1C=NC=N1)C1=CC=C(Cl)C=C1Cl
1
–
7
of
7
results
2
Penconazole, TRC
CAS: 66246-88-6 Molecular Formula: C13 H15 Cl2 N3 Molecular Weight (g/mol): 284.18 Synonym: 1H-1,2,4-Triazole, 1-[2-(2,4-dichlorophenyl)pentyl]-,1-[2-(2,4-Dichlorophenyl)pentyl]-1H-1,2,4-triazole,(±)-Penconazole,CGA 71818,Erluejunzuo,Ofir,Omnex,Omnex WP 10,Penconazole,Pine gel,Regal gel,Topas,Topas (fungicide),Topaze IUPAC Name: 1-[2-(2,4-dichlorophenyl)pentyl]-1,2,4-triazole SMILES: CCCC(Cn1cncn1)c2ccc(Cl)cc2Cl