missing translation for 'onlineSavingsMsg'
Learn More

Pefabloc™ SC (4-(2-Aminoethyl)-benzolsulfonylfluorid-hydrochloride), Millipore™

Product Code. 10760522
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10760522 100 mg 100mg
10423781 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10760522 Supplier Millipore Supplier No. 1248390100
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AEBSF for biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30827-99-7

Specifications

Product Type Inhibitor
Synonym Pefabloc™ SC (4-(2-Aminoethyl)-benzolsulfonylfluorid-hydrochloride), (4-(2-aminoetyl)-benzenesulfonyl fluoride hydrochloride), Pefabloc™ SC (4-(2-Aminoethyl)-benzolsulfonylfluorid-hydrochloride)
Form Solid
Quantity 100 mg
Molecular Weight (g/mol) 239.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.