Learn More
Novus Biologicals™ Peanut Agglutinin [Cy5]
Product Code. 18371468 Shop All Bio Techne Products
Ansicht ändern
Click to view available options
Quantity:
1 mg
Packungsgröße:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
18371468 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18371468 Lieferant Novus Biologicals™ Lieferanten-Nr. CL1075NB
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Spezifikation
Antigen Peanut Agglutinin
Concentration 1 mg/mL
Quantity 1 mg
Content And Storage Store at 4°C in the dark.
For Use With (Application) A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37C and centrifuge before use. The recommended concentration range for use is 5-20 μg/mL.
Buffer 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM CaCl2, proprietary stabilizer