Learn More
PDZD3 293T Cell Overexpression Lysate (Denatured), Abnova
Product Code. 16120107
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16120107 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16120107 Supplier Abnova Supplier No. H00079849T01.100uL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Western Blotting
Specifications
Cell Line 293T
For Use With (Application) Western Blot
Description PDZD3 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_024791
Molecular Weight (g/mol) 54.12
pH Range 6.8
Plasmid pCMV-PDZD3 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-PDZD3 antibody (H00079849-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL PDZD3 293T Cell Transient Overexpression Lys
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Mercaptoethanol