missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDCL2 Polyclonal specifically detects PDCL2 in Human/Mouse/Rat samples. It is validated for ELISA/Western Blot.
Specifications
Specifications
| Antigen | PDCL2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GCPHLP, phosducin-like 2, phosducin-like protein 2 |
| Host Species | Rabbit |
| Immunogen | PDCL2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human PDCL2. The immunogen is located within the last 50 amino acids of PDCL2. Amino Acid Squence: NTSIHDDSDSSNSDNDT |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?