Learn More
Patchouli Alcohol, TRC
Product Code. 30011641
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011641 25 mg 25mg
30006151 250 mg 250mg
30006935 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011641 Supplier Toronto Research Chemicals Supplier No. TRCP20620025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5986-55-0
Molecular Formula C15 H26 O
Molecular Weight (g/mol) 222.37
Synonym 1,6-Methanonaphthalen-1(2H)-ol, octahydro-4,8a,9,9-tetramethyl-, (1R,4S,4aS,6R,8aS)-, 1,6-Methanonaphthalen-1(2H)-ol, octahydro-4,8a,9,9-tetramethyl-, [1R-(1α,4β,4aα,6β,8aα)]-, 1,6-Methanonaphthalen-1β(2H)-ol, 3,4,4aβ,5,6β,7,8,8a-octahydro-4α,8aβ,9,9-tetramethyl- (8CI), Patchouli alcohol (6CI,7CI), (-)-Patchouli alcohol, (-)-Patchoulol, (1R,4S,4aS,6R,8aS)-Octahydro-4,8a,9,9-tetramethyl-1,6-methanonaphthalen-1(2H)-ol, Patchoulol
SMILES C[C@H]1CC[C@@]2(O)C(C)(C)[C@@H]3CC[C@@]2(C)[C@H]1C3
Specifikationer
Quantity 25 mg
InChI Formula InChI=1S/C15H26O/c1-10-5-8-15(16)13(2,3)11-6-7-14(15,4)12(10)9-11/h10-12,16H,5-9H2,1-4H3/t10-,11+,12-,14-,15+/m0/s1
Formula Weight 222.1984
Chemical Name or Material Patchouli Alcohol
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.