CAS RN 5986-55-0
CAS RN 5986-55-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Patchouli Alcohol, TRC
CAS: 5986-55-0 Molecular Formula: C15 H26 O Molecular Weight (g/mol): 222.37 Synonym: 1,6-Methanonaphthalen-1(2H)-ol, octahydro-4,8a,9,9-tetramethyl-, (1R,4S,4aS,6R,8aS)-,1,6-Methanonaphthalen-1(2H)-ol, octahydro-4,8a,9,9-tetramethyl-, [1R-(1α,4β,4aα,6β,8aα)]-,1,6-Methanonaphthalen-1β(2H)-ol, 3,4,4aβ,5,6β,7,8,8a-octahydro-4α,8aβ,9,9-tetramethyl- (8CI),Patchouli alcohol (6CI,7CI),(-)-Patchouli alcohol,(-)-Patchoulol,(1R,4S,4aS,6R,8aS)-Octahydro-4,8a,9,9-tetramethyl-1,6-methanonaphthalen-1(2H)-ol,Patchoulol SMILES: C[C@H]1CC[C@@]2(O)C(C)(C)[C@@H]3CC[C@@]2(C)[C@H]1C3