missing translation for 'onlineSavingsMsg'
Learn More

Palmitoleic Acid Methyl Ester, MP Biomedicals™

Product Code. 11419371 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419371 1 g 1g
11409371 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419371 Supplier MP Biomedicals Supplier No. 0215604891
  • ONLINE EXCLUSIVE PRICE
    299.00 EUR
    284.55 EUR / 1g
    Save 14.45 EUR
    4% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Palmitoleic Acid Methyl Ester, MP Biomedicals is a component in the preparation of biodiesel fuels.

Chemical Identifiers

CAS 1120-25-8
Molecular Formula C17H32O2
Molecular Weight (g/mol) 268.44
MDL Number MFCD00042911
InChI Key IZFGRAGOVZCUFB-HJWRWDBZSA-N
Synonym methyl palmitoleate, z-methyl hexadec-9-enoate, methyl z-hexadec-9-enoate, methyl palmitoleinate, unii-35695qdb9f, methyl 9z-hexadec-9-enoate, 9-hexadecenoic acid, methyl ester, z, methyl cis-9-hexadecenoate, methyl z-9-hexadecenoate, methyl cis-hexadec-9-enoate
PubChem CID 643801
ChEBI CHEBI:84156
IUPAC Name methyl (9Z)-hexadec-9-enoate
SMILES CCCCCC\C=C/CCCCCCCC(=O)OC

Specifications

Melting Point -0.5°C to +0.5°C (literature)
Density 0.875g/mL at 25°C (Lit.)
Boiling Point 180°C to 183°C (1 mmHg)(literature)
Refractive Index n20/D 1.451 (Lit.)
Quantity 1 g
Formula Weight 268.44
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.