accessibility menu, dialog, popup

UserName

CAS RN 1120-25-8

IUPAC Name: methyl (9Z)-hexadec-9-enoate
Synonyms: methyl (9Z)-hexadec-9-enoate
Molecular Weight (g/mol): 268.44
Molecular Formula: C17H32O2
InChi Key: IZFGRAGOVZCUFB-HJWRWDBZSA-N
SMILES: CCCCCC\C=C/CCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)