Learn More
Alfa Aesar™ Palladium(II) chloride, solution, Pd 20-25% w/w (cont. Pd), Thermo Scientific™
Product Code. 11470241
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470241 1 g 1g
11460241 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11470241 Supplier Alfa Aesar™ Supplier No. 040019.NH
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
IUPAC Name dichloropalladium
SMILES [Cl-].[Cl-].[Pd++]
Specifications
Color Brown
UN Number UN3264
Concentration Pd 20 to 25% w/w (cont. Pd)
Solubility Information Soluble in aqueous metal halides,water,acetone,alcohol,hydrobromic acid and hydrochloric acid.
Formula Weight 177.31
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Palladium(II) chloride Solution