accessibility menu, dialog, popup

UserName

ClClPd

CAS RN 7647-10-1

IUPAC Name: palladium(2+) dichloride
Synonyms: palladium(II) chloride
Molecular Weight (g/mol): 177.32
Molecular Formula: Cl2Pd
InChi Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++]
Molecular Weight (g/mol) 
  • (27)
  • (3)
Quantity 
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (9)
  • (2)
Grade 
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (6)
  • (14)
  • (3)
  • (4)
  • (27)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (4)
  • (9)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)