missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Oncostatin M/OSM |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | 15 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | MGC20461, oncostatin M, oncostatin-M |
| Host Species | Rabbit |
| Immunogen | Recombinant Human Oncostatin M/OSM protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?