Learn More
Tocris Bioscience™ Oltipraz
Description
Oltipraz is a Nrf2 activator; activates Nrf, which binds to antioxidant response elements (AREs) and induces Phase-I and -II drug-metabolizing enzymes including CYP2B and NQO1. Also increases mRNA levels of multiple hepatic xenobiotic transporters. Elevates expression of genes encoding anti-oxidant and multidrug resistance-associated proteins. Displays chemopreventive activity.
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-5-(2-pyrazinyl)-3H-1,2-dithiole-3-thione |
| CAS | 64224-21-1 |
| Quantity | 50 mg |
| Target | Nrf2 Modulators |
| Molecular Formula | C8H6N2S3 |
| Formula Weight | 226.34 |
| Purity | >99% |