Learn More
o-Toluidine Chloride, TRC
Product Code. 30136546
Change view
Click to view available options
Quantity:
50 mg
5 g
25 g
Unit Size:
25g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136546 25 g 25g
30005088 5 g 5g
30012123 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136546 Supplier Toronto Research Chemicals Supplier No. TRCT53621525G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 636-21-5
Molecular Formula C7 H9 N . Cl H
Molecular Weight (g/mol) 143.61
Synonym o-Toluidine Hydrochloride, Benzenamine, 2-methyl-, hydrochloride (1:1) (ACI), Benzenamine, 2-methyl-, hydrochloride (9CI), o-Toluidine, hydrochloride (8CI), 2-Methylaniline hydrochloride, 2-Methylbenzenamine hydrochloride, 2-Methylphenylammonium chloride, o-Methylaniline hydrochloride, o-Toluidinium chloride
IUPAC Name 2-methylaniline;hydrochloride
SMILES Cl.Cc1ccccc1N
Specifications
Quantity 25 g
InChI Formula InChI=1S/C7H9N.ClH/c1-6-4-2-3-5-7(6)8;/h2-5H,8H2,1H3;1H
Formula Weight 143.0502
Chemical Name or Material 2-methylbenzenamine hydrochloride
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.