accessibility menu, dialog, popup

UserName

CAS RN 636-21-5

HClCH3H2N

CAS RN 636-21-5

IUPAC Name: hydrogen 2-methylaniline chloride
Synonyms: hydrogen O-toluidine chloride
Molecular Weight (g/mol): 143.61
Molecular Formula: C7H10ClN
InChi Key: OGVXWEOZQMAAIM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results