Learn More
O-tert-Butyl-L-tyrosine, 99%, Thermo Scientific Chemicals
Product Code. 11319779
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319779 1 g 1g
11329779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11319779 Supplier Thermo Scientific Alfa Aesar Supplier No. B22575.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18822-59-8
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD00037202
InChI Key SNZIFNXFAFKRKT-NSHDSACASA-N
Synonym h-tyr tbu-oh, o-tert-butyl-l-tyrosine, o-t-butyl-l-tyrosine, h-l-tyr tbu-oh, tyrosine, o-1,1-dimethylethyl, s-2-amino-3-4-tert-butoxy phenyl propanoic acid, 2s-2-amino-3-4-tert-butoxy phenyl propanoic acid, o-tert.butyl-l-tyrosine, o-t-butyl-tyrosine, tyr but
PubChem CID 7408092
IUPAC Name (2S)-2-amino-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@H]([NH3+])C([O-])=O)C=C1
Specifications
Melting Point ∼205°C (decomposition)
Quantity 1 g
Formula Weight 237.3
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material O-tert-Butyl-L-tyrosine