accessibility menu, dialog, popup

UserName

(S)ONH2OHOH3CCH3CH3

CAS RN 18822-59-8

IUPAC Name: (2S)-2-azaniumyl-3-[4-(tert-butoxy)phenyl]propanoate
Synonyms: (2S)-2-ammonio-3-[4-(tert-butoxy)phenyl]propanoate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: SNZIFNXFAFKRKT-NSHDSACASA-N
SMILES: CC(C)(C)OC1=CC=C(C[C@H]([NH3+])C([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)