missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Bovine, Drosophila, Fish, Hamster, Human, Mouse, Pig, Primate, Rat, Virus, Xenopus species samples. It is validated for ChIP Assay, Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications
Specifications
| Antigen | O-GlcNAc |
| Applications | ChIP Assay, Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | RL2 |
| Conjugate | PE-Cyanine5.5 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | GlcNAc, O-linked N-acetylglucosamine |
| Host Species | Mouse |
| Immunogen | Pore complex-lamina fraction purified from rat liver nuclear envelopes. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?