missing translation for 'onlineSavingsMsg'
Learn More

O-Aminophenol Sulfate, TRC

Product Code. 30137402
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137402 500 mg 500mg
30135989 100 mg 100mg
17983455 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137402 Supplier Toronto Research Chemicals Supplier No. TRCA576865500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67845-79-8
Molecular Formula 2 C6 H7 N O . H2 S O4
Molecular Weight (g/mol) 316.33
Synonym o-Aminophenol sulfate
IUPAC Name 2-aminophenol;sulfuric acid
SMILES Nc1ccccc1O.Nc2ccccc2O.OS(=O)(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2C6H7NO.H2O4S/c2*7-5-3-1-2-4-6(5)8;1-5(2,3)4/h2*1-4,8H,7H2;(H2,1,2,3,4)
Formula Weight 316.0729
Chemical Name or Material di-2-Aminophenol sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.