CAS RN 67845-79-8
CAS RN 67845-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
O-Aminophenol Sulfate, TRC
CAS: 67845-79-8 Molecular Formula: 2 C6 H7 N O . H2 S O4 Molecular Weight (g/mol): 316.33 Synonym: o-Aminophenol sulfate,o-Aminophenol sulfate IUPAC Name: 2-aminophenol;sulfuric acid SMILES: Nc1ccccc1O.Nc2ccccc2O.OS(=O)(=O)O