missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | NTB-A/SLAMF6/CD352 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 40 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 |
| Host Species | Rabbit |
| Immunogen | Recombinant Mouse NTB-A/SLAMF6/CD352 Protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?