Learn More
NP-AHD, TRC
Product Code. 30135624
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135624 250 mg 250mg
30012015 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135624 Supplier Toronto Research Chemicals Supplier No. TRCN894900250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 623145-57-3
Molecular Formula C10 H8 N4 O4
Molecular Weight (g/mol) 248.19
Synonym 1-[[(2-Nitrophenyl)methylene]amino]-2,4-imidazolidinedione
IUPAC Name 1-[(E)-(2-nitrophenyl)methylideneamino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)c1ccccc1\C=N\N2CC(=O)NC2=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C10H8N4O4/c15-9-6-13(10(16)12-9)11-5-7-3-1-2-4-8(7)14(17)18/h1-5H,6H2,(H,12,15,16)/b11-5+
Formula Weight 248.0546
Percent Purity >95%
Chemical Name or Material 2-NP-AHD
Grade HPLC