CAS RN 623145-57-3
CAS RN 623145-57-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
NP-AHD, TRC
CAS: 623145-57-3 Molecular Formula: C10 H8 N4 O4 Molecular Weight (g/mol): 248.19 Synonym: 1-[[(2-Nitrophenyl)methylene]amino]-2,4-imidazolidinedione IUPAC Name: 1-[(E)-(2-nitrophenyl)methylideneamino]imidazolidine-2,4-dione SMILES: [O-][N+](=O)c1ccccc1\C=N\N2CC(=O)NC2=O