missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ UCK2 Protein

Product Code. 18767243 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767243 0.1 mg 0.1mg
18215041 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18767243 Supplier Novus Biologicals™ Supplier No. NBP1508880.1mg
  • ONLINE EXCLUSIVE PRICE
    428.00 EUR
    404.25 EUR / 0.1mg
    Save 23.75 EUR
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-261 of Human UCK2 The Recombinant Human UCK2 Protein is derived from E. coli. The Recombinant Human UCK2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT, 100mM NaCl, 0.1mM PMSF, 1mM EDTA
Molecular Weight (g/mol) 30.3kDa
Quantity 0.1 mg
Regulatory Status RUO
Conjugate Unconjugated
Purity or Quality Grade >90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.